missing translation for 'onlineSavingsMsg'
Learn More

2,4-Di(tert-pentyl)phenoxyacetic acid, 98%, Thermo Scientific™

Product Code. 11408477
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13402-96-5
Molecular Formula C18H27O3
Molecular Weight (g/mol) 291.41
MDL Number MFCD00128799
InChI Key QXQMENSTZKYZCE-UHFFFAOYSA-M
Synonym 2-2,4-di-tert-pentylphenoxy acetic acid, 2,4-di tert-amyl phenoxyacetic acid, 2,4-di-tert-pentylphenoxy acetic acid, 2,4-di tert-pentyl phenoxyacetic acid, 2,4-di-tert-pentylphenoxyacetic acid, 2-2,4-bis 2-methylbutan-2-yl phenoxy acetic acid, acetic acid, 2,4-bis 1,1-dimethylpropyl phenoxy, 2,4-bis 1,1-dimethylpropyl phenoxy acetic acid, acetic acid, 2-2,4-bis 1,1-dimethylpropyl phenoxy
PubChem CID 83411
IUPAC Name 2-[2,4-bis(2-methylbutan-2-yl)phenoxy]acetic acid
SMILES CCC(C)(C)C1=CC=C(OCC([O-])=O)C(=C1)C(C)(C)CC

Specifications

Melting Point 125°C to 127°C
Quantity 25 g
Formula Weight 292.42
Percent Purity 98%
Chemical Name or Material 2,4-Di(tert-pentyl)phenoxyacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'