missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dibromo-6-fluoroaniline, 97%, Thermo Scientific™

Product Code. 10642623
Click to view available options
Quantity:
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 141474-37-5
Molecular Formula C6H4Br2FN
Molecular Weight (g/mol) 268.91
MDL Number MFCD00042230
InChI Key YJLXEKFYZIBUPJ-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dibromo-6-fluoro, 2,4-dibromo-6-fluorobenzenamine, 2,4-dibromo-6-fluoro-phenylamine, 2,4-dibromo-6-fluoro aniline, pubchem3442, intermediates-zcf02078, ksc494o1h, 2,4-dibromo-6-fluoro-aniline, acmc-1c814, 4,6-dibromo-2-fluorophenylamine
PubChem CID 2736765
IUPAC Name 2,4-dibromo-6-fluoroaniline
SMILES NC1=C(F)C=C(Br)C=C1Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 268.91
Percent Purity 97%
Chemical Name or Material 2, 4-Dibromo-6-fluoroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'