Learn More
2,4-Dichloro-1-(4-nitrophenoxy)benzene, 97%, Thermo Scientific™
£7.00 - £90.00
Chemical Identifiers
CAS | 1836-75-5 |
---|---|
Molecular Formula | C12H7Cl2NO3 |
Molecular Weight (g/mol) | 284.092 |
MDL Number | MFCD00128026 |
InChI Key | XITQUSLLOSKDTB-UHFFFAOYSA-N |
Synonym | nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen |
PubChem CID | 15787 |
IUPAC Name | 2,4-dichloro-1-(4-nitrophenoxy)benzene |
SMILES | C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10723063
|
Thermo Scientific Maybridge
RDP00053DA |
1 g | Amber Glass Bottle | ||||||
10202792
|
Thermo Scientific Maybridge
RDP00053EA |
10 g | Amber Glass Bottle | ||||||
10387244
|
Thermo Scientific Maybridge
RDP00053EB |
25 g | Amber Glass Bottle | ||||||
Description
Chemical Identifiers
1836-75-5 | |
284.092 | |
XITQUSLLOSKDTB-UHFFFAOYSA-N | |
15787 | |
C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl |
C12H7Cl2NO3 | |
MFCD00128026 | |
nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen | |
2,4-dichloro-1-(4-nitrophenoxy)benzene |
Specifications
1836-75-5 | |
1.3000g/mL | |
C12H7Cl2NO3 | |
MFCD00128026 | |
XITQUSLLOSKDTB-UHFFFAOYSA-N | |
2,4-dichloro-1-(4-nitrophenoxy)benzene | |
15787 | |
97% |
97.0 | |
97% | |
Cl2C6H3OC6H4NO2 | |
nitrofen, 2,4-dichloro-1-4-nitrophenoxy benzene, nitrochlor, nitrophen, niclofen, nitrophene, trizilin, mezotox, nitrafen, nitraphen | |
C1=CC(=CC=C1[N+](=O)[O-])OC2=C(C=C(C=C2)Cl)Cl | |
284.092 | |
284.1 | |
2, 4-Dichloro-1-(4-nitrophenoxy)benzene |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Very toxic to aquatic life with long lasting effects.
May cause cancer.
May damage the unborn child.
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger