missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-1-iodobenzene, 97%, Thermo Scientific™

£33.40 - £54.10

Chemical Identifiers

CAS 29898-32-6
Molecular Formula C6H3Cl2I
Molecular Weight (g/mol) 272.894
InChI Key ZQKJCBDCOGLKCQ-UHFFFAOYSA-N
Synonym 1,3-dichloro-4-iodobenzene, 2,4-dichloroiodobenzene, benzene, 2,4-dichloro-1-iodo, 1-iodo-2,4-dichlorobenzene, pubchem3692, 2,4-dichlor-jodbenzol, acmc-209hbu, 1,3-dichloro4-iodobenzene, 1,3-dichloro 4-iodobenzene, 2,4-dichloro-1-iodo-benzene
PubChem CID 96864
IUPAC Name 2,4-dichloro-1-iodobenzene
SMILES C1=CC(=C(C=C1Cl)Cl)I
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10767495
View Documents
Thermo Scientific Maybridge
TL00185EE
50 g
£33.40
50g
Please to purchase this item. Need a web account? Register with us today!
10566334
missing translation for 'viewDocuments'
Thermo Scientific Maybridge
TL00185FA
100 g
£54.10
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

29898-32-6
272.894
1,3-dichloro-4-iodobenzene, 2,4-dichloroiodobenzene, benzene, 2,4-dichloro-1-iodo, 1-iodo-2,4-dichlorobenzene, pubchem3692, 2,4-dichlor-jodbenzol, acmc-209hbu, 1,3-dichloro4-iodobenzene, 1,3-dichloro 4-iodobenzene, 2,4-dichloro-1-iodo-benzene
2,4-dichloro-1-iodobenzene
C6H3Cl2I
ZQKJCBDCOGLKCQ-UHFFFAOYSA-N
96864
C1=CC(=C(C=C1Cl)Cl)I

Specifications

29898-32-6
2.0100g/mL
C6H3Cl2I
ZQKJCBDCOGLKCQ-UHFFFAOYSA-N
2,4-dichloro-1-iodobenzene
96864
97%
97.0
97%
1,3-dichloro-4-iodobenzene, 2,4-dichloroiodobenzene, benzene, 2,4-dichloro-1-iodo, 1-iodo-2,4-dichlorobenzene, pubchem3692, 2,4-dichlor-jodbenzol, acmc-209hbu, 1,3-dichloro4-iodobenzene, 1,3-dichloro 4-iodobenzene, 2,4-dichloro-1-iodo-benzene
C1=CC(=C(C=C1Cl)Cl)I
272.894
272.9
2, 4-Dichloro-1-iodobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Certificates