missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-3-ethyl-6-nitrophenol, 98%, Thermo Scientific™
£19.95 - £58.70
Chemical Identifiers
CAS | 99817-36-4 |
---|---|
Molecular Formula | C8H6Cl2NO3 |
Molecular Weight (g/mol) | 235.04 |
MDL Number | MFCD00270764 |
InChI Key | YTVCECQSAPGJBB-UHFFFAOYSA-M |
Synonym | 2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol |
PubChem CID | 7020339 |
IUPAC Name | 2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate |
SMILES | CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15214854
|
Thermo Scientific Acros
450760010 |
1 g | Glass bottle | ||||||
15204854
|
Thermo Scientific Acros
450760050 |
5 g | Glass bottle |
£58.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
99817-36-4 | |
235.04 | |
YTVCECQSAPGJBB-UHFFFAOYSA-M | |
7020339 | |
CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O |
C8H6Cl2NO3 | |
MFCD00270764 | |
2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol | |
2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate |
Specifications
99817-36-4 | |
97.5% min. (HPLC) | |
MFCD00270764 | |
Solubility in water: insoluble. Other solubilities: soluble in methanol | |
CCC1=C(Cl)C=C(C([O-])=C1Cl)[N+]([O-])=O | |
235.04 | |
236.05 | |
2, 4-Dichloro-3-ethyl-6-nitrophenol |
49.0°C to 51.0°C | |
C8H6Cl2NO3 | |
2,4-dichloro-3-ethyl-6-nitrophenol, 2,4-dichloro-3-ethyl-6-nitro-phenol, phenol, 2,4-dichloro-3-ethyl-6-nitro, 2-nitro-4,6-dichloro-5-ethylphenol, 2,4-dichloro-3-ethyl-6-nitrochloride, pubchem20442, acmc-209se5, ksc486m4b, 2,4-bis chloranyl-3-ethyl-6-nitro-phenol | |
YTVCECQSAPGJBB-UHFFFAOYSA-M | |
2,4-dichloro-3-ethyl-6-nitrobenzen-1-olate | |
7020339 | |
98% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED
GHS Signal Word: Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2,4-Dichloro-3-ethyl-6-nitrophenol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only