missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloro-5-sulfamoylbenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11440438
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2736-23-4
Molecular Formula C7H5Cl2NO4S
Molecular Weight (g/mol) 270.08
MDL Number MFCD00007931
InChI Key ZSHHRBYVHTVRFK-UHFFFAOYSA-N
Synonym lasamide, 5-aminosulfonyl-2,4-dichlorobenzoic acid, 2,4-dichloro-5-sulphamoylbenzoic acid, 2,4-dichloro-5-sulfamoyl-benzoic acid, unii-leg53tf0sn, 2,4-dichloro-5-sulfamylbenzoic acid, benzoic acid, 5-aminosulfonyl-2,4-dichloro, leg53tf0sn, 3-sulfamoyl-4,6-dichlorobenzoic acid, benzoic acid, 2,4-dichloro-5-sulfamoyl
PubChem CID 17655
IUPAC Name 2,4-dichloro-5-sulfamoylbenzoic acid
SMILES C1=C(C(=CC(=C1S(=O)(=O)N)Cl)Cl)C(=O)O

Specifications

Melting Point 230°C to 232°C (decomposition)
Quantity 250 g
Formula Weight 270.09
Percent Purity 98%
Chemical Name or Material 2,4-Dichloro-5-sulfamoylbenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'