missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dichloro-7H-pyrrolo[2,3-d]pyrimidine, 95%, Thermo Scientific™
£199.56 - £522.17
Chemical Identifiers
CAS | 90213-66-4 |
---|---|
Molecular Formula | C6H3Cl2N3 |
Molecular Weight (g/mol) | 188.02 |
InChI Key | GHXBPCSSQOKKGB-UHFFFAOYSA-N |
Synonym | 2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l |
PubChem CID | 14116871 |
IUPAC Name | 2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine |
SMILES | C1=CNC2=C1C(=NC(=N2)Cl)Cl |
Description
Chemical Identifiers
90213-66-4 | |
188.02 | |
2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l | |
2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine |
C6H3Cl2N3 | |
GHXBPCSSQOKKGB-UHFFFAOYSA-N | |
14116871 | |
C1=CNC2=C1C(=NC(=N2)Cl)Cl |
Specifications
90213-66-4 | |
94% min. (HPLC) | |
2,4-dichloro-7h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro-1h-pyrrolo 2,3-d pyrimidine, 2,6-dichloro-7-deazapurine, 2,4-dichloropyrrolo 2,3-d pyrimidine, 2,4-dichloro-7h-pyrrolo2,3-dpyrimidine, 1h-pyrrolo 2,3-d pyrimidine, 2,4-dichloro, pubchem14941, acmc-209unt, tofacitinib intermediate ii, ksc486m2l | |
C1=CNC2=C1C(=NC(=N2)Cl)Cl | |
188.02 | |
188.02 | |
2, 4-Dichloro-7H-pyrrolo[2, 3-d]pyrimidine |
250°C to 252°C | |
C6H3Cl2N3 | |
GHXBPCSSQOKKGB-UHFFFAOYSA-N | |
2,4-dichloro-7H-pyrrolo[2,3-d]pyrimidine | |
14116871 | |
95% |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2,4-Dichloro-7H-pyrrolo¢2,3-d!pyrimidine, 95%
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only