Learn More
2,4-Dichloroaniline, 98%, Thermo Scientific Chemicals
£18.10 - £433.22
Chemical Identifiers
CAS | 554-00-7 |
---|---|
Molecular Formula | C6H5Cl2N |
Molecular Weight (g/mol) | 162.02 |
MDL Number | MFCD00007661 |
InChI Key | KQCMTOWTPBNWDB-UHFFFAOYSA-N |
Synonym | 2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german |
PubChem CID | 11123 |
ChEBI | CHEBI:46635 |
IUPAC Name | 2,4-dichloroaniline |
SMILES | C1=CC(=C(C=C1Cl)Cl)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10438490
|
Thermo Scientific Acros
150390250 |
25 g | Glass bottle |
£18.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10347370
|
Thermo Scientific Acros
150392500 |
250 g | Glass bottle |
£60.80
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10541781
|
Thermo Scientific Acros
150390010 |
1 kg | Glass bottle |
£189.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13428368
|
Thermo Scientific Acros
150390025 |
2.5 kg | Plastic bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
554-00-7 | |
162.02 | |
KQCMTOWTPBNWDB-UHFFFAOYSA-N | |
11123 | |
2,4-dichloroaniline |
C6H5Cl2N | |
MFCD00007661 | |
2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german | |
CHEBI:46635 | |
C1=CC(=C(C=C1Cl)Cl)N |
Specifications
554-00-7 | |
100.0 | |
White to Beige | |
>110°C | |
97.5% min. (GC) | |
Cl2C6H3NH2 | |
12,621 | |
Solubility in water: insoluble | |
C1=CC(=C(C=C1Cl)Cl)N | |
162.02 | |
CHEBI:46635 | |
98% | |
2, 4-Dichloroaniline, 98% |
97.5 | |
59°C to 63°C | |
242°C | |
Authentic | |
C6H5Cl2N | |
MFCD00007661 | |
2,4-dichlorobenzenamine, benzenamine, 2,4-dichloro, 2,4-dichloranilin, o,p-dichloroaniline, aniline, 2,4-dichloro, 2,4-dca, 2,4-dichlorophenylamine, 2,4-dichloro-phenylamine, unii-19ae42m6ws, 2,4-dichloranilin german | |
KQCMTOWTPBNWDB-UHFFFAOYSA-N | |
2,4-dichloroaniline | |
11123 | |
162.02 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-057-8
missing translation for 'rtecsNumber' : BX2600000
missing translation for 'tsca' : TSCA
Recommended Storage : Product darkens on storage
- 2,4-Dichloroaniline
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H301-Toxic if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only