Learn More
2,4-Dichlorobenzyl mercaptan, 98+%, Thermo Scientific Chemicals
£32.40 - £109.00
Chemical Identifiers
CAS | 59293-67-3 |
---|---|
Molecular Formula | C7H6Cl2S |
Molecular Weight (g/mol) | 193.085 |
MDL Number | MFCD00039650 |
InChI Key | ZSPXTTVUJDSRNJ-UHFFFAOYSA-N |
Synonym | 2,4-dichlorobenzyl mercaptan, 2,4-dichlorophenyl methanethiol, 2,4-dichlorobenzylmercaptan, 2,4-dichlorobenzenemethanethiol, 2,4-dichlorotoluene-alpha-thiol, benzenemethanethiol, 2,4-dichloro, 2,4-dichloro-alpha-toluenethiol, acmc-1b9kz, d0sz9f, 2,4-dichlorothiobenzyl alcohol |
PubChem CID | 95756 |
IUPAC Name | (2,4-dichlorophenyl)methanethiol |
SMILES | C1=CC(=C(C=C1Cl)Cl)CS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11461587
|
Thermo Scientific Alfa Aesar
B24999.06 |
5 g |
£32.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11471587
|
Thermo Scientific Alfa Aesar
B24999.14 |
25 g |
£109.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
59293-67-3 | |
193.085 | |
ZSPXTTVUJDSRNJ-UHFFFAOYSA-N | |
95756 | |
C1=CC(=C(C=C1Cl)Cl)CS |
C7H6Cl2S | |
MFCD00039650 | |
2,4-dichlorobenzyl mercaptan, 2,4-dichlorophenyl methanethiol, 2,4-dichlorobenzylmercaptan, 2,4-dichlorobenzenemethanethiol, 2,4-dichlorotoluene-alpha-thiol, benzenemethanethiol, 2,4-dichloro, 2,4-dichloro-alpha-toluenethiol, acmc-1b9kz, d0sz9f, 2,4-dichlorothiobenzyl alcohol | |
(2,4-dichlorophenyl)methanethiol |
Spezifikation
59293-67-3 | |
150°C to 151°C (12 mmHg) | |
C7H6Cl2S | |
MFCD00039650 | |
509602 | |
2,4-dichlorobenzyl mercaptan, 2,4-dichlorophenyl methanethiol, 2,4-dichlorobenzylmercaptan, 2,4-dichlorobenzenemethanethiol, 2,4-dichlorotoluene-alpha-thiol, benzenemethanethiol, 2,4-dichloro, 2,4-dichloro-alpha-toluenethiol, acmc-1b9kz, d0sz9f, 2,4-dichlorothiobenzyl alcohol | |
C1=CC(=C(C=C1Cl)Cl)CS | |
193.085 | |
193.1 | |
2,4-Dichlorobenzyl mercaptan |
1.36 | |
>100°C (212°F) | |
1.6045 | |
UN2810 | |
Air Sensitive | |
ZSPXTTVUJDSRNJ-UHFFFAOYSA-N | |
(2,4-dichlorophenyl)methanethiol | |
95756 | |
≥98% |
Sicherheit und Handhabung
GHS H Statement
H331-H302-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P311-P330-P332+P313-P362-P501c
H302-H315-H319-H331-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 261-687-2
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only