missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichlorophenol, 99%

Product Code. 10427540 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 10427540

Brand: Thermo Scientific Acros 147721000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 120-83-2 | C6H4Cl2O | 163.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-83-2
Molecular Formula C6H4Cl2O
Molecular Weight (g/mol) 163.00
MDL Number MFCD00002169
InChI Key HFZWRUODUSTPEG-UHFFFAOYSA-N
Synonym 4,6-dichlorophenol, phenol, 2,4-dichloro, 2,4-dcp, 2,4-dichloro-phenol, 1-hydroxy-2,4-dichlorobenzene, rcra waste number u081, 2,4-dichlorohydroxybenzene, ccris 657, rcra waste no. u081, 1,3-dichloro-4-hydroxybenzene
PubChem CID 8449
ChEBI CHEBI:16738
IUPAC Name 2,4-dichlorophenol
SMILES OC1=CC=C(Cl)C=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 41°C to 44°C
Color White to Beige
Boiling Point 209°C to 210°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3OH
Quantity 100 g
Beilstein 06, 189
Merck Index 15, 3082
Solubility Information Solubility in water: 4.5g/L in water (20°C). Other solubilities: soluble in carbon tetrachloride,diethyl ether,,benzene,chloroform,ethanol,aromatic solvents,,acetone and sodium hydroxide
Formula Weight 163
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 2, 4-Dichlorophenol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Dichlorophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.