missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dichloroquinazoline, 98%, Thermo Scientific Chemicals

£36.68 - £139.70

Chemical Identifiers

CAS 607-68-1
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.03
MDL Number MFCD00091950
InChI Key TUQSVSYUEBNNKQ-UHFFFAOYSA-N
Synonym 2,4-dichloro-quinazoline, quinazoline, 2,4-dichloro, dichloroquinazoline, 2,4-dichlorchinazolin, pubchem9668, 2,4-dichloroquinazol, 2,4dichloroquinazoline, acmc-209mlw, 2,4-dichloroquinazoline;, 2, 4-dichloro-quinazoline
PubChem CID 252886
IUPAC Name 2,4-dichloroquinazoline
SMILES ClC1=NC(Cl)=C2C=CC=CC2=N1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15537529
View Documents
Thermo Scientific Acros
460730050
5 g
£36.68
5g
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
15547529
View Documents
Thermo Scientific Acros
460730250
25 g
£139.70
25g
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

607-68-1
199.03
TUQSVSYUEBNNKQ-UHFFFAOYSA-N
252886
ClC1=NC(Cl)=C2C=CC=CC2=N1
C8H4Cl2N2
MFCD00091950
2,4-dichloro-quinazoline, quinazoline, 2,4-dichloro, dichloroquinazoline, 2,4-dichlorchinazolin, pubchem9668, 2,4-dichloroquinazol, 2,4dichloroquinazoline, acmc-209mlw, 2,4-dichloroquinazoline;, 2, 4-dichloro-quinazoline
2,4-dichloroquinazoline

Specifications

607-68-1
Authentic
C8H4Cl2N2
2,4-dichloro-quinazoline, quinazoline, 2,4-dichloro, dichloroquinazoline, 2,4-dichlorchinazolin, pubchem9668, 2,4-dichloroquinazol, 2,4dichloroquinazoline, acmc-209mlw, 2,4-dichloroquinazoline;, 2, 4-dichloro-quinazoline
ClC1=NC(Cl)=C2C=CC=CC2=N1
199.03
199.04
2,4-Dichloroquinazoline
115.0°C to 121.0°C
97.5% min. (GC)
MFCD00091950
TUQSVSYUEBNNKQ-UHFFFAOYSA-N
2,4-dichloroquinazoline
252886
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2,4-Dichloroquinazoline, 98%

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only