missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-1-iodobenzene, 99%, Thermo Scientific Chemicals

Product Code. 11419722
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2265-93-2
Molecular Formula C6H3F2I
Molecular Weight (g/mol) 239.99
MDL Number MFCD00040151
InChI Key YKLDMAPEGQYZRT-UHFFFAOYSA-N
Synonym 2,4-difluoroiodobenzene, 1,3-difluoro-4-iodobenzene, 2,4-difluoro-1-iodobenze, benzene, 2,4-difluoro-1-iodo, 2,4-difluoro iodobenzene, 2,4-difluoro-1-iodo-benzene, pubchem3463, 2,4-difluoro-iodobenzene, acmc-1cq0v, intermediates-zcf02607
PubChem CID 137519
IUPAC Name 2,4-difluoro-1-iodobenzene
SMILES FC1=CC=C(I)C(F)=C1

Specifications

Density 2.006
Boiling Point 175°C to 176°C
Flash Point 67°C (152°F)
Refractive Index 1.558
Quantity 250 g
Beilstein 2081078
Sensitivity Light sensitive
Formula Weight 239.99
Percent Purity 99%
Chemical Name or Material 2,4-Difluoro-1-iodobenzene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'