missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-3-methoxybenzyl bromide, 97%, Thermo Scientific™

Product Code. 11362499
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11362499

Brand: Thermo Scientific Alfa Aesar H26625.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 886499-17-8
Molecular Formula C8H7BrF2O
Molecular Weight (g/mol) 237.044
MDL Number MFCD04115936
InChI Key DXDNJSHUSXGESC-UHFFFAOYSA-N
Synonym 2,4-difluoro-3-methoxybenzyl bromide, 1-bromomethyl-2,4-difluoro-3-methoxybenzene, 4-bromomethyl-1,3-difluoro-2-methoxybenzene
PubChem CID 3833566
IUPAC Name 1-(bromomethyl)-2,4-difluoro-3-methoxybenzene
SMILES COC1=C(C=CC(=C1F)CBr)F

Specifications

Refractive Index 1.5275
Quantity 1 g
UN Number UN3265
Formula Weight 237.04
Percent Purity 97%
Chemical Name or Material 2,4-Difluoro-3-methoxybenzyl bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.