missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluoro-5-nitrobenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 15435367
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 153775-33-8
Molecular Formula C7H2F2NO4
Molecular Weight (g/mol) 202.09
MDL Number MFCD04115713
InChI Key MDFSGDMPHMKKGB-UHFFFAOYSA-M
PubChem CID 2782822
IUPAC Name 2,4-difluoro-5-nitrobenzoate
SMILES [O-]C(=O)C1=CC(=C(F)C=C1F)[N+]([O-])=O

Specifications

Melting Point 125°C to 133°C
Quantity 1 g
Synonym 2,4-difluoro-5-nitrobenzoic acid, benzoic acid, 2,4-difluoro-5-nitro, benzoic acid,2,4-difluoro-5-nitro, 2,4-difluoro-5-nitro-benzoic acid, acmc-209d9v, ksc527s6t, 2,4-difluoro-5-nitrobenzoicacid, buttpark 156\01-93
Formula Weight 203.1
Percent Purity 97%
Chemical Name or Material 2,4-Difluoro-5-nitrobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'