Learn More
2',4'-Difluoroacetophenone, 98%, Thermo Scientific Chemicals
£19.20 - £67.30
Chemical Identifiers
CAS | 364-83-0 |
---|---|
Molecular Formula | C8H6F2O |
Molecular Weight (g/mol) | 156.132 |
MDL Number | MFCD00151261 |
InChI Key | QEWHNJPLPZOEKU-UHFFFAOYSA-N |
Synonym | 2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213 |
PubChem CID | 67770 |
IUPAC Name | 1-(2,4-difluorophenyl)ethanone |
SMILES | CC(=O)C1=C(C=C(C=C1)F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11465453
|
Thermo Scientific Alfa Aesar
A13420.06 |
5 g | |||||||
11475453
|
Thermo Scientific Alfa Aesar
A13420.14 |
25 g |
£67.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
364-83-0 | |
156.132 | |
QEWHNJPLPZOEKU-UHFFFAOYSA-N | |
67770 | |
CC(=O)C1=C(C=C(C=C1)F)F |
C8H6F2O | |
MFCD00151261 | |
2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213 | |
1-(2,4-difluorophenyl)ethanone |
Specifications
364-83-0 | |
80°C to 81°C (25 mmHg) | |
Characteristic | |
1.488 | |
1940741 | |
QEWHNJPLPZOEKU-UHFFFAOYSA-N | |
1-(2,4-difluorophenyl)ethanone | |
67770 | |
98% |
1.232 | |
66°C (150°F) | |
C8H6F2O | |
MFCD00151261 | |
2',4'-difluoroacetophenone, 1-2,4-difluorophenyl ethanone, 2,4-difluoroacetophenone, 1-2,4-difluorophenyl ethan-1-one, ethanone, 1-2,4-difluorophenyl, 1-2,4-difluoro-phenyl-ethanone, 1-acetyl-2,4-difluorobenzene, acetophenone, 2',4'-difluoro, 1-2,4-difluorphenyl ethanon, pubchem4213 | |
CC(=O)C1=C(C=C(C=C1)F)F | |
156.132 | |
156.13 | |
2',4'-Difluoroacetophenone |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 206-667-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only