missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzophenone, 97+%, Thermo Scientific™

Product Code. 11426236
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-35-5
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.203
MDL Number MFCD00000331
InChI Key FRHMSTHFVFIPCO-UHFFFAOYSA-N
Synonym 2,4-difluorobenzophenone, 2,4-difluorophenyl phenyl methanone, 2,4-difluorophenyl-phenylmethanone, methanone, 2,4-difluorophenyl phenyl, 2,4-difluorophenyl phenyl ketone, fr cf dvr, 2,4-bis fluoranyl phenyl-phenyl-methanone, 2,4-difluorobenzo-phenone, frhmsthfvfipco-uhfffaoysa, 2,4-difluorophenyl-phenyl-methanone
PubChem CID 570834
IUPAC Name (2,4-difluorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=C(C=C(C=C2)F)F

Specifications

Density 1.261
Boiling Point 119°C to 120°C (2 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.569
Quantity 25 g
Beilstein 6596192
Formula Weight 218.2
Percent Purity ≥97%
Chemical Name or Material 2,4-Difluorobenzophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'