missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Difluorobenzoyl chloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
2,4-Difluorobenzoyl chloride is used as a pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from water/ moisture, oxidizing agents, bases.
Specifications
Specifications
Density | 1.437 |
Boiling Point | 96°C to 98°C (15 mmHg) |
Flash Point | 82°C (179°F) |
Refractive Index | 1.5145 |
Quantity | 25 g |
UN Number | UN3265 |
Beilstein | 4800566 |
Sensitivity | Moisture sensitive |
Synonym | 2,4-difluorobenzoylchloride, benzoyl chloride, 2,4-difluoro, benzoyl chloride,2,4-difluoro, acmc-1bfet, 2,4difluorobenzoylchloride, 2,4-difluorobezoyl chloride, 2,4 difluorobenzoyl chloride, 2,4-difluoro benzoylchloride, 2,4-difluoro-benzoylchloride, 2,4-difluoro-benzoyl chloride |
Solubility Information | Reacts with water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'