Learn More
2,4-Difluorophenyl isocyanate, 98+%, Thermo Scientific Chemicals
£32.00 - £262.00
Chemical Identifiers
CAS | 59025-55-7 |
---|---|
Molecular Formula | C7H3F2NO |
Molecular Weight (g/mol) | 155.10 |
MDL Number | MFCD00001997 |
InChI Key | HNENEALJPWJWJY-UHFFFAOYSA-N |
Synonym | 2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k |
PubChem CID | 593258 |
IUPAC Name | 2,4-difluoro-1-isocyanatobenzene |
SMILES | FC1=CC=C(N=C=O)C(F)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499593
|
Thermo Scientific Alfa Aesar
A12415.06 |
5 g |
£32.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409603
|
Thermo Scientific Alfa Aesar
A12415.14 |
25 g |
£94.70
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419603
|
Thermo Scientific Alfa Aesar
A12415.22 |
100 g |
£262.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59025-55-7 | |
155.10 | |
HNENEALJPWJWJY-UHFFFAOYSA-N | |
593258 | |
FC1=CC=C(N=C=O)C(F)=C1 |
C7H3F2NO | |
MFCD00001997 | |
2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k | |
2,4-difluoro-1-isocyanatobenzene |
Specifications
59025-55-7 | |
64°C to 66°C (10 mmHg) | |
C7H3F2NO | |
MFCD00001997 | |
2831577 | |
2,4-difluorophenyl isocyanate, 2,4-difluorophenylisocyanate, benzene, 2,4-difluoro-1-isocyanato, isocyanic acid 2,4-difluorophenyl ester, 2,4-difluoro-1-isocyanato-benzene, 2,4-difluoroisocyanatobenzene, timtec-bb sbb006537, pubchem4453, 2,4-diflurophenylisocyanate, acmc-1b06k | |
FC1=CC=C(N=C=O)C(F)=C1 | |
155.10 | |
155.11 | |
2,4-Difluorophenyl isocyanate |
1.31 | |
50°C (122°F) | |
1.492 | |
UN3080 | |
Moisture sensitive | |
HNENEALJPWJWJY-UHFFFAOYSA-N | |
2,4-difluoro-1-isocyanatobenzene | |
593258 | |
≥98% |
Safety and Handling
GHS H Statement
H300-H330-H334-H226-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: II
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only