missing translation for 'onlineSavingsMsg'
Learn More
2,4-Difluorophenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals Product Code.: 11399126

2,4-Difluorophenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals

Product Code. 11399126
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11399126

Brand: Thermo Scientific Alfa Aesar L20266.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79784-36-4
Molecular Formula C8H4F2O2
Molecular Weight (g/mol) 170.12
MDL Number MFCD04038287
InChI Key AZBQYELTMQNVST-UHFFFAOYSA-N
Synonym 2,4-difluorophenylglyoxal hydrate, 2-2,4-difluorophenyl-2-oxoacetaldehyde hydrate, 2,4-difluorophenylglyoxal monohydrate, benzeneacetaldehyde,2,4-difluoro-a-oxo, 2-2,4-difluorophenyl-2-oxoethanal, hydrate, 2-2,4-bis fluoranyl phenyl-2-oxidanylidene-ethanal hydrate
PubChem CID 2782300
IUPAC Name 2-(2,4-difluorophenyl)-2-oxoacetaldehyde;hydrate
SMILES FC1=CC(F)=C(C=C1)C(=O)C=O

Specifications

Melting Point 51°C to 54°C
Quantity 1 g
Beilstein 4387465
Solubility Information Soluble in water.
Formula Weight 170.12 (Anhydrous)
Percent Purity 95%
Assay (dry wt.)
Chemical Name or Material 2,4-Difluorophenylglyoxal hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Difluorophenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.