missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Difluoropropiophenone, 98%, Thermo Scientific™

Product Code. 11449902
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85068-30-0
Molecular Formula C9H8F2O
Molecular Weight (g/mol) 170.16
MDL Number MFCD00015507
InChI Key UZWOADNMVRRYDE-UHFFFAOYSA-N
Synonym 2',4'-difluoropropiophenone, 1-2,4-difluorophenyl propan-1-one, 2,4-difluoropropiophenone, 1-propanone, 1-2,4-difluorophenyl, 2,4-defluoropropiophenone, 1-2,4-difluorophenyl-1-propanone, 1-2.4-difluoro-phenyl-propan-1-one, acmc-209q3o, ksc654a2r
PubChem CID 123586
IUPAC Name 1-(2,4-difluorophenyl)propan-1-one
SMILES CCC(=O)C1=CC=C(F)C=C1F

Specifications

Density 1.193
Boiling Point 51°C to 52°C (1.5 mmHg)
Flash Point 76°C (168°F)
Refractive Index 1.485
Quantity 25 g
Beilstein 7633370
Formula Weight 170.16
Percent Purity 98%
Chemical Name or Material 2',4'-Difluoropropiophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'