missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2',4'-Difluoropropiophenone, 98%, Thermo Scientific™
£28.30 - £345.00
Chemical Identifiers
CAS | 85068-30-0 |
---|---|
Molecular Formula | C9H8F2O |
Molecular Weight (g/mol) | 170.16 |
MDL Number | MFCD00015507 |
InChI Key | UZWOADNMVRRYDE-UHFFFAOYSA-N |
Synonym | 2',4'-difluoropropiophenone, 1-2,4-difluorophenyl propan-1-one, 2,4-difluoropropiophenone, 1-propanone, 1-2,4-difluorophenyl, 2,4-defluoropropiophenone, 1-2,4-difluorophenyl-1-propanone, 1-2.4-difluoro-phenyl-propan-1-one, acmc-209q3o, ksc654a2r |
PubChem CID | 123586 |
IUPAC Name | 1-(2,4-difluorophenyl)propan-1-one |
SMILES | CCC(=O)C1=CC=C(F)C=C1F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439902
|
Thermo Scientific Alfa Aesar
A12032.06 |
5 g | |||||||
11449902
|
Thermo Scientific Alfa Aesar
A12032.14 |
25 g |
£117.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459902
|
Thermo Scientific Alfa Aesar
A12032.22 |
100 g | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
85068-30-0 | |
170.16 | |
UZWOADNMVRRYDE-UHFFFAOYSA-N | |
123586 | |
CCC(=O)C1=CC=C(F)C=C1F |
C9H8F2O | |
MFCD00015507 | |
2',4'-difluoropropiophenone, 1-2,4-difluorophenyl propan-1-one, 2,4-difluoropropiophenone, 1-propanone, 1-2,4-difluorophenyl, 2,4-defluoropropiophenone, 1-2,4-difluorophenyl-1-propanone, 1-2.4-difluoro-phenyl-propan-1-one, acmc-209q3o, ksc654a2r | |
1-(2,4-difluorophenyl)propan-1-one |
Specifications
85068-30-0 | |
51°C to 52°C (1.5 mmHg) | |
C9H8F2O | |
MFCD00015507 | |
2',4'-difluoropropiophenone, 1-2,4-difluorophenyl propan-1-one, 2,4-difluoropropiophenone, 1-propanone, 1-2,4-difluorophenyl, 2,4-defluoropropiophenone, 1-2,4-difluorophenyl-1-propanone, 1-2.4-difluoro-phenyl-propan-1-one, acmc-209q3o, ksc654a2r | |
CCC(=O)C1=CC=C(F)C=C1F | |
170.16 | |
170.16 | |
2',4'-Difluoropropiophenone |
1.193 | |
76°C (168°F) | |
1.485 | |
7633370 | |
UZWOADNMVRRYDE-UHFFFAOYSA-N | |
1-(2,4-difluorophenyl)propan-1-one | |
123586 | |
98% |
Safety and Handling
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
missing translation for 'einecsNumber' : 285-291-4
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures