missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dihydroxybenzoic acid, 97%, Thermo Scientific Chemicals

Product Code. 11460024
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Dihydroxybenzoic acid is used in Intermediates, Plasticizers, Plastic and Rubber Products, a organic building block.

Solubility
Soluble in water(70g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-86-1
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002451
InChI Key UIAFKZKHHVMJGS-UHFFFAOYSA-N
Synonym beta-resorcylic acid, p-hydroxysalicylic acid, 4-carboxyresorcinol, 4-hydroxysalicylic acid, benzoic acid, 2,4-dihydroxy, beta-resorcinolic acid, 2,4-dhba, resorcylic acid, beta, b-resorcylic acid, 2,4-dihydroxy-benzoic acid
PubChem CID 1491
IUPAC Name 2,4-dihydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)O)C(=O)O

Specifications

Melting Point ∼226°C (decomposition)
Quantity 100 g
Beilstein 1946213
Merck Index 14,8157
Solubility Information Soluble in water(70g/L).
Formula Weight 154.12
Percent Purity 97%
Chemical Name or Material 2,4-Dihydroxybenzoic acid
compliance-icons
Product Identifier
  • 2,4-Dihydroxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2,4-Dihydroxybenzoicacid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'