Learn More
2,4-Dimethoxybenzoyl chloride, 96%, Thermo Scientific™
£17.78 - £191.00
Chemical Identifiers
CAS | 39828-35-8 |
---|---|
Molecular Formula | C9H9ClO3 |
Molecular Weight (g/mol) | 200.618 |
MDL Number | MFCD00017527 |
InChI Key | KVIUXRJCBBXEGJ-UHFFFAOYSA-N |
Synonym | 2,4-dimethoxybenzoylchloride, acmc-20aocn, 2, 4-dimethoxybenzoyl chloride, 2,4-dimethoxy benzoyl chloride, 2,4-dimethoxy-benzoyl chloride, benzoyl chloride,2,4-dimethoxy, 2,4-dimethoxybenzoic acid chloride, 2,4-bis methyloxy benzoyl chloride, 2,4-dimethoxy-1-benzenecarbonyl chloride, benzoyl chloride, 2,4-dimethoxy-9ci |
PubChem CID | 2734719 |
IUPAC Name | 2,4-dimethoxybenzoyl chloride |
SMILES | COC1=CC(=C(C=C1)C(=O)Cl)OC |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
39828-35-8 | |
200.618 | |
KVIUXRJCBBXEGJ-UHFFFAOYSA-N | |
2734719 | |
COC1=CC(=C(C=C1)C(=O)Cl)OC |
C9H9ClO3 | |
MFCD00017527 | |
2,4-dimethoxybenzoylchloride, acmc-20aocn, 2, 4-dimethoxybenzoyl chloride, 2,4-dimethoxy benzoyl chloride, 2,4-dimethoxy-benzoyl chloride, benzoyl chloride,2,4-dimethoxy, 2,4-dimethoxybenzoic acid chloride, 2,4-bis methyloxy benzoyl chloride, 2,4-dimethoxy-1-benzenecarbonyl chloride, benzoyl chloride, 2,4-dimethoxy-9ci | |
2,4-dimethoxybenzoyl chloride |
Specifications
39828-35-8 | |
C9H9ClO3 | |
UN3261 | |
2,4-dimethoxybenzoylchloride, acmc-20aocn, 2, 4-dimethoxybenzoyl chloride, 2,4-dimethoxy benzoyl chloride, 2,4-dimethoxy-benzoyl chloride, benzoyl chloride,2,4-dimethoxy, 2,4-dimethoxybenzoic acid chloride, 2,4-bis methyloxy benzoyl chloride, 2,4-dimethoxy-1-benzenecarbonyl chloride, benzoyl chloride, 2,4-dimethoxy-9ci | |
COC1=CC(=C(C=C1)C(=O)Cl)OC | |
200.618 | |
200.62 | |
2,4-Dimethoxybenzoyl chloride |
57°C to 61°C | |
MFCD00017527 | |
Moisture sensitive | |
KVIUXRJCBBXEGJ-UHFFFAOYSA-N | |
2,4-dimethoxybenzoyl chloride | |
2734719 | |
96% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312+H332-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold