missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethoxybenzyl alcohol, 99%, Thermo Scientific Chemicals
£81.60 - £258.00
Chemical Identifiers
CAS | 7314-44-5 |
---|---|
Molecular Formula | C9H12O3 |
Molecular Weight (g/mol) | 168.19 |
MDL Number | MFCD00004614 |
InChI Key | RNKOUSCCPHSCFE-UHFFFAOYSA-N |
Synonym | 2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol |
PubChem CID | 81750 |
IUPAC Name | (2,4-dimethoxyphenyl)methanol |
SMILES | COC1=CC(=C(C=C1)CO)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10523921
|
Thermo Scientific Acros
170460250 |
25 g | Glass bottle |
£81.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10157613
|
Thermo Scientific Acros
170461000 |
100 g | Glass bottle |
£258.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7314-44-5 | |
168.19 | |
RNKOUSCCPHSCFE-UHFFFAOYSA-N | |
81750 | |
COC1=CC(=C(C=C1)CO)OC |
C9H12O3 | |
MFCD00004614 | |
2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol | |
(2,4-dimethoxyphenyl)methanol |
Specifications
7314-44-5 | |
100.0 | |
177.0°C to 179.0°C (10.0 mmHg) | |
Authentic | |
C9H12O3 | |
1.5450 to 1.5480 | |
06, I, 550 | |
RNKOUSCCPHSCFE-UHFFFAOYSA-N | |
(2,4-dimethoxyphenyl)methanol | |
81750 | |
99% | |
2, 4-Dimethoxybenzyl alcohol |
98.5 | |
37.0°C to 40.0°C | |
>110°C | |
98.5% min. (GC) | |
(CH3O)2C6H3CH2OH | |
MFCD00004614 | |
2,4-dimethoxybenzyl alcohol, 2,4-dimethoxyphenyl methanol, benzenemethanol, 2,4-dimethoxy, 2,4-dimethoxybenzylalcohol, 1-hydroxymethyl-2,4-dimethoxybenzene, 2,4-dimethoxyphenyl methan-1-ol, pubchem7454, acmc-209opp, 2,4-dimethoxy-benzylalcohol, 2,4-dimethoxybenzyl-alcohol | |
COC1=CC(=C(C=C1)CO)OC | |
168.19 | |
168.19 | |
Low Melting Solid |
Safety and Handling
missing translation for 'einecsNumber' : 230-775-2
RUO – Research Use Only