Learn More
2,4-Dimethoxybenzylamine, 98%, Thermo Scientific Chemicals
£37.00 - £112.00
Chemical Identifiers
CAS | 20781-20-8 |
---|---|
Molecular Formula | C9H13NO2 |
Molecular Weight (g/mol) | 167.208 |
MDL Number | MFCD00052393 |
InChI Key | QOWBXWFYRXSBAS-UHFFFAOYSA-N |
PubChem CID | 597250 |
IUPAC Name | (2,4-dimethoxyphenyl)methanamine |
SMILES | COC1=CC(=C(C=C1)CN)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11476250
|
Thermo Scientific Alfa Aesar
H27140.06 |
5 g |
£37.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11486250
|
Thermo Scientific Alfa Aesar
H27140.14 |
25 g |
£112.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20781-20-8 | |
167.208 | |
QOWBXWFYRXSBAS-UHFFFAOYSA-N | |
(2,4-dimethoxyphenyl)methanamine |
C9H13NO2 | |
MFCD00052393 | |
597250 | |
COC1=CC(=C(C=C1)CN)OC |
Specifications
20781-20-8 | |
140°C (1 mmHg) | |
C9H13NO2 | |
MFCD00052393 | |
Air Sensitive | |
QOWBXWFYRXSBAS-UHFFFAOYSA-N | |
(2,4-dimethoxyphenyl)methanamine | |
597250 | |
98% |
1.113 | |
>110°C (230°F) | |
1.549 | |
UN2735 | |
2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine | |
COC1=CC(=C(C=C1)CN)OC | |
167.208 | |
167.21 | |
2,4-Dimethoxybenzylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only