missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dimethoxyphenyl isocyanate, 97%, Thermo Scientific Chemicals Product Code.: 11332257

2,4-Dimethoxyphenyl isocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11332257
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11332257

Brand: Thermo Scientific Alfa Aesar L09762.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84370-87-6
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.175
MDL Number MFCD00002005
InChI Key WRAZHLRMDRTLOZ-UHFFFAOYSA-N
Synonym 2,4-dimethoxyphenyl isocyanate, 2,4-dimethoxyphenylisocyanate, benzene, 1-isocyanato-2,4-dimethoxy, 2,4-dimethoxybenzenisocyanate, acmc-20aocl, 1-isocyanato-2,4-dimethoxy-benzene, #, benzene,1-isocyanato-2,4-dimethoxy, 1-isocyanato-2,4-bis methyloxy benzene
PubChem CID 601637
IUPAC Name 1-isocyanato-2,4-dimethoxybenzene
SMILES COC1=CC(=C(C=C1)N=C=O)OC

Specifications

Melting Point 31°C to 32°C
Boiling Point 139°C to 141°C (11 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5475
Quantity 5 g
UN Number UN2811
Beilstein 1106649
Sensitivity Moisture sensitive
Formula Weight 179.18
Percent Purity 97%
Chemical Name or Material 2,4-Dimethoxyphenyl isocyanate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251761 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberL09762.06,L09762.03
Lot Number10247016 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberL09762.06,L09762.03
Lot Number10239243 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL09762.06,L09762.03
Lot Number10221845 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL09762.06,L09762.03
Lot Number10193781 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL09762.06,L09762.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dimethoxyphenyl isocyanate, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.