missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethyl-1-pentene, 99%, Thermo Scientific Chemicals

£120.60 - £420.09

Chemical Identifiers

CAS 2213-32-3
Molecular Formula C7H14
Molecular Weight (g/mol) 98.189
MDL Number MFCD00039854
InChI Key LXQPBCHJNIOMQU-UHFFFAOYSA-N
Synonym 2,4-dimethyl-1-pentene, 1-pentene, 2,4-dimethyl, acmc-1ch0z, pentene, 2,4-dimethyl, ch3 2chch2c ch3 =ch2, 1-pentene, 2,4-dimethyl-8ci 9ci
PubChem CID 16657
IUPAC Name 2,4-dimethylpent-1-ene
SMILES CC(C)CC(=C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11400451
SDS View Documents
Thermo Scientific Alfa Aesar
B20002.88
2.5 g
£120.60
2.50g
Please to purchase this item. Need a web account? Register with us today!
11410451
SDS View Documents
Thermo Scientific Alfa Aesar
B20002.09
10 g
£420.09
10g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

missing translation for 'einecsNumber' : 218-665-2

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dimethyl-1-pentene, 99%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.