missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-dimethyl-3-furaldehyde, Thermo Scientific™
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 124.14 |
Percent Purity | 95% |
Chemical Name or Material | 2, 4-dimethyl-3-furaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'