missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethyl-3-furoic acid, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 140.14 |
Percent Purity | 97% |
Chemical Name or Material | 2, 4-Dimethyl-3-furoic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'