missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dimethylaniline, 99%, Thermo Scientific Chemicals

Product Code. 10736191
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-68-1
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00007738
InChI Key CZZZABOKJQXEBO-UHFFFAOYSA-N
Synonym 2,4-xylidine, 2,4-dimethyl aniline, m-xylidine, m-4-xylidine, 2,4-dimethylphenylamine, benzenamine, 2,4-dimethyl, 2-methyl-p-toluidine, 4-methyl-o-toluidine, 1-amino-2,4-dimethylbenzene, 4-amino-1,3-xylene
PubChem CID 7250
ChEBI CHEBI:27840
IUPAC Name 2,4-dimethylaniline
SMILES CC1=CC(=C(C=C1)N)C

Specifications

Melting Point 16.0°C
CAS Min % 98.5
CAS Max % 100.0
Density 0.9800g/mL
Boiling Point 218.0°C
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5580 to 1.5600
Assay Percent Range 98.5% min. (GC)
Specific Gravity 0.98
Linear Formula (CH3)2C6H3NH2
Beilstein 12, 1111
Merck Index 15, 10282
Solubility Information Solubility in water: 5g/L (20°C). Other solubilities: soluble in ethanol, ether and benzene
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,4-Dimethylaniline, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,4-Dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'