Learn More
2,4-Dimethylthiazole, 99%, Thermo Scientific Chemicals
£22.60 - £171.00
Chemical Identifiers
CAS | 541-58-2 |
---|---|
Molecular Formula | C5H7NS |
Molecular Weight (g/mol) | 113.18 |
MDL Number | MFCD00014509 |
InChI Key | OBSLLHNATPQFMJ-UHFFFAOYSA-N |
Synonym | 2,4-dimethylthiazole, thiazole, 2,4-dimethyl, 2,4-methylthiazole, 2,4-dimethyl thiazole, unii-lht97038oa, 2,4-dimethyl-thiazole, dimethylthiazol, zlchem 533, 2,4 dimethylthiazole, 2.4-dimethylthiazole |
PubChem CID | 10934 |
IUPAC Name | 2,4-dimethyl-1,3-thiazole |
SMILES | CC1=NC(C)=CS1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460154
|
Thermo Scientific Alfa Aesar
A13604.06 |
5 g |
£22.60
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470154
|
Thermo Scientific Alfa Aesar
A13604.14 |
25 g |
£73.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480154
|
Thermo Scientific Alfa Aesar
A13604.22 |
100 g |
£171.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
541-58-2 | |
113.18 | |
OBSLLHNATPQFMJ-UHFFFAOYSA-N | |
10934 | |
CC1=NC(C)=CS1 |
C5H7NS | |
MFCD00014509 | |
2,4-dimethylthiazole, thiazole, 2,4-dimethyl, 2,4-methylthiazole, 2,4-dimethyl thiazole, unii-lht97038oa, 2,4-dimethyl-thiazole, dimethylthiazol, zlchem 533, 2,4 dimethylthiazole, 2.4-dimethylthiazole | |
2,4-dimethyl-1,3-thiazole |
Specifications
541-58-2 | |
42°C (107°F) | |
C5H7NS | |
UN1993 | |
2,4-dimethylthiazole, thiazole, 2,4-dimethyl, 2,4-methylthiazole, 2,4-dimethyl thiazole, unii-lht97038oa, 2,4-dimethyl-thiazole, dimethylthiazol, zlchem 533, 2,4 dimethylthiazole, 2.4-dimethylthiazole | |
OBSLLHNATPQFMJ-UHFFFAOYSA-N | |
2,4-dimethyl-1,3-thiazole | |
10934 | |
99% |
145°C to 147°C | |
Characteristic | |
MFCD00014509 | |
106414 | |
Slightly soluble in water | |
CC1=NC(C)=CS1 | |
113.18 | |
113.18 | |
2,4-Dimethylthiazole |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
missing translation for 'rtecsNumber' : XJ4375000
missing translation for 'tsca' : TSCA
RUO – Research Use Only