missing translation for 'onlineSavingsMsg'
Learn More
2,4-Dinitro-1-(trifluoromethoxy)benzene, 98%, Thermo Scientific™ Product Code.: 11400447

2,4-Dinitro-1-(trifluoromethoxy)benzene, 98%, Thermo Scientific™

Product Code. 11400447
2.5 g
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy

Product Code. 11400447

Brand: Thermo Scientific Alfa Aesar B21790.88

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 655-07-2
Molecular Formula C7H3F3N2O5
Molecular Weight (g/mol) 252.11
MDL Number MFCD00042268
InChI Key KYUUZCUXYXNPFO-UHFFFAOYSA-N
Synonym 2,4-dinitro-1-trifluoromethoxy benzene, 2,4-dinitro trifluoromethoxy benzene, 2,4-dinitrotrifluoroanisole, 2,4-dinitrotrifluoromethoxybenzene, 2,4-dinitro-1-trifluoromethoxy-benzene, acmc-20aoc3, 2,4-dinitro trifluoroanisole, a,a,a-trifluoro-2,4-dinitro-anisole, 1-trifluoromethoxy-2,4-dinitrobenzene, benzene,2,4-dinitro-1-trifluoromethoxy
PubChem CID 2737131
IUPAC Name 2,4-dinitro-1-(trifluoromethoxy)benzene
SMILES [O-][N+](=O)C1=CC=C(OC(F)(F)F)C(=C1)[N+]([O-])=O

Specifications

Melting Point -21°C
Density 1.623
Boiling Point 273°C to 274°C
Flash Point 154°C (309°F)
Quantity 2.5 g
UN Number UN2927
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 252.11
Percent Purity 98%
Chemical Name or Material 2,4-Dinitro-1-(trifluoromethoxy)benzene

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10150502 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10201436 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,4-Dinitro-1-(trifluoromethoxy)benzene, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.