missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dinitrobenzenesulfonic acid sodium salt, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
Specifications
Specifications
Quantity | 25 g |
Beilstein | 6676807 |
Solubility Information | Soluble in water and methanol. |
Formula Weight | 270.15 |
Percent Purity | 97% |
Chemical Name or Material | 2,4-Dinitrobenzenesulfonic acid sodium salt |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction