missing translation for 'onlineSavingsMsg'
Learn More

2,4-Dinitrophenylhydrazine, moist solid, contains min. 30% water, Thermo Scientific Chemicals

Chemical Identifiers

CAS 119-26-6, 7732-18-5
Molecular Formula C6H6N4O4
MDL Number MFCD00007578
InChI Key HORQAOAYAYGIBM-UHFFFAOYSA-N
Synonym 2,4-dinitrophenyl hydrazine, 2,4-dnph, hydrazine, 2,4-dinitrophenyl, 1-hydrazino-2,4-dinitrobenzene, dnph, brady's reagent, 2,4-dinitrofenylhydrazin, unii-1n39kd7qpj, ccris 3140, 2,4-dnp hydrazine
PubChem CID 3772977
ChEBI CHEBI:66932
IUPAC Name (2,4-dinitrophenyl)hydrazine
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

119-26-6, 7732-18-5
MFCD00007578
2,4-dinitrophenyl hydrazine, 2,4-dnph, hydrazine, 2,4-dinitrophenyl, 1-hydrazino-2,4-dinitrobenzene, dnph, brady's reagent, 2,4-dinitrofenylhydrazin, unii-1n39kd7qpj, ccris 3140, 2,4-dnp hydrazine
CHEBI:66932
C6H6N4O4
HORQAOAYAYGIBM-UHFFFAOYSA-N
3772977
(2,4-dinitrophenyl)hydrazine

Specifications

119-26-6, 7732-18-5
Brown-Orange to Yellow
C6H6N4O4
MFCD00007578
01,330; 02,176
2,4-dinitrophenyl hydrazine, 2,4-dnph, hydrazine, 2,4-dinitrophenyl, 1-hydrazino-2,4-dinitrobenzene, dnph, brady's reagent, 2,4-dinitrofenylhydrazin, unii-1n39kd7qpj, ccris 3140, 2,4-dnp hydrazine
C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NN
198.138
CHEBI:66932
≥98.0% (HPLC)
2, 4-Dinitrophenylhydrazine
198°C to 204°C
Authentic
(O2N)2C6H3NHNH2
15, 489
15, 3311
HORQAOAYAYGIBM-UHFFFAOYSA-N
(2,4-dinitrophenyl)hydrazine
3772977
198.14
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable solid.
Suspected of causing cancer.
Harmful if swallowed.
Explosive when dry.

GHS P Statement:
Use personal protective equipment as required.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

missing translation for 'einecsNumber' : 204-309-3

missing translation for 'rtecsNumber' : MV3325000

missing translation for 'tsca' : TSCA

Product Identifier
  • 2,4-Dinitrophenylhydrazine, moist solid, contains min. 30% water

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 1
  • Acute toxicity Category 4

Hazard Statement
  • H228-Flammable solid.
  • H302-Harmful if swallowed.
  • EUH001-Explosive when dry.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P264-Wash thoroughly after handling.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only