missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diphenyl-1,3-thiazole-5-carbaldehyde, ≥95%, Thermo Scientific™

Product Code. 10331293
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10331293

Brand: Thermo Scientific Maybridge CC42304DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 864068-85-9
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Molecular Formula C16H11NOS
MDL Number MFCD08060521
Quantity 1 g
Synonym 2,4-diphenylthiazole-5-carbaldehyde, 5-thiazolecarboxaldehyde,2,4-diphenyl
InChI Key QCAWTHNKWALNJR-UHFFFAOYSA-N
SMILES O=CC1=C(N=C(S1)C1=CC=CC=C1)C1=CC=CC=C1
IUPAC Name 2,4-diphenyl-1,3-thiazole-5-carbaldehyde
Molecular Weight (g/mol) 265.33
PubChem CID 7537553
Formula Weight 265.33
Percent Purity 95%
Chemical Name or Material 2, 4-Diphenyl-1, 3-thiazole-5-carbaldehyde
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.