Learn More
2-(4-Fluorophenyl)ethylamine, 97%, Thermo Scientific Chemicals
£24.90 - £102.00
Chemical Identifiers
CAS | 1583-88-6 |
---|---|
Molecular Formula | C8H10FN |
Molecular Weight (g/mol) | 139.173 |
MDL Number | MFCD00134208 |
InChI Key | CKLFJWXRWIQYOC-UHFFFAOYSA-N |
Synonym | 4-fluorophenethylamine, 2-4-fluorophenyl ethanamine, 2-4-fluoro-phenyl-ethylamine, p-fluorophenethylamine, 4-fluorophenylethylamine, 2-4-fluorophenyl ethylamine, 2-4-fluorophenyl ethan-1-amine, benzeneethanamine, 4-fluoro, p-fluorophenyl ethylamine, 2-4-fluorophenyl-ethylamine |
PubChem CID | 4653 |
IUPAC Name | 2-(4-fluorophenyl)ethanamine |
SMILES | C1=CC(=CC=C1CCN)F |
Description
2-(4-Fluorophenyl)ethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives. It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(4-Fluorophenyl)ethylamine may be employed as nucleophile in the synthesis of 2-amino-4-arylpyrimidine derivatives. It is suitable for use in the preparation of ortho-metalated primary phenethylamines having electron-releasing and electron-withdrawing groups on the aromatic ring, leading to complexes containing six membered palladacycles.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
Chemical Identifiers
1583-88-6 | |
139.173 | |
CKLFJWXRWIQYOC-UHFFFAOYSA-N | |
4653 | |
C1=CC(=CC=C1CCN)F |
C8H10FN | |
MFCD00134208 | |
4-fluorophenethylamine, 2-4-fluorophenyl ethanamine, 2-4-fluoro-phenyl-ethylamine, p-fluorophenethylamine, 4-fluorophenylethylamine, 2-4-fluorophenyl ethylamine, 2-4-fluorophenyl ethan-1-amine, benzeneethanamine, 4-fluoro, p-fluorophenyl ethylamine, 2-4-fluorophenyl-ethylamine | |
2-(4-fluorophenyl)ethanamine |
Specifications
1583-88-6 | |
1.061 g/mL | |
79°C (174°F) | |
1.507 | |
UN2922 | |
Slightly soluble in water. | |
C1=CC(=CC=C1CCN)F | |
139.173 | |
139.17 | |
Liquid |
Yellow | |
50°C to 52°C | |
C8H10FN | |
MFCD00134208 | |
4-fluorophenethylamine, 2-4-fluorophenyl ethanamine, 2-4-fluoro-phenyl-ethylamine, p-fluorophenethylamine, 4-fluorophenylethylamine, 2-4-fluorophenyl ethylamine, 2-4-fluorophenyl ethan-1-amine, benzeneethanamine, 4-fluoro, p-fluorophenyl ethylamine, 2-4-fluorophenyl-ethylamine | |
CKLFJWXRWIQYOC-UHFFFAOYSA-N | |
2-(4-fluorophenyl)ethanamine | |
4653 | |
97% | |
2-(4-Fluorophenyl)ethylamine |
Safety and Handling
GHS H Statement
H300-H310-H330-H314-H318-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 216-435-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only