missing translation for 'onlineSavingsMsg'
Learn More

2-[(4-Fluorophenyl)sulfonyl]-N'-hydroxyethanimidamide, 97%, Thermo Scientific™

Product Code. 10447432
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175203-76-6
Molecular Formula C8H9FN2O3S
Molecular Weight (g/mol) 232.229
MDL Number MFCD00205110
InChI Key STPCRDRACVTHTE-UHFFFAOYSA-N
Synonym 2-4-fluorophenyl sulfonyl-n'-hydroxyethanimidamide, 2-4-fluorobenzenesulfonyl acetamide oxime, 2-4-fluorobenzenesulphonyl acetamide oxime, 2-4-fluorophenyl sulfonyl-n-hydroxyacetimidamide, 2-4-fluorophenyl sulfonyl-n-hydroxyethanimidamide, 1-4-fluorophenyl sulfonyl-2-hydroxyimino eth-2-ylamine, 2-4-fluorobenzenesulphonyl acetamidoxime, 2-4-fluorobenzenesulfonyl-n'-hydroxyethanimidamide, z-2-4-fluorobenzenesulfonyl-n'-hydroxyethanimidamide, 2-4-fluorophenylsulfonyl-n-hydroxy-acetamidine
PubChem CID 9580384
IUPAC Name 2-(4-fluorophenyl)sulfonyl-N'-hydroxyethanimidamide
SMILES C1=CC(=CC=C1F)S(=O)(=O)CC(=NO)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula FC6H4SO2CH2C(=NOH)NH2
Quantity 1 g
Formula Weight 232.23
Percent Purity 97%
Chemical Name or Material 2-[(4-Fluorophenyl)sulfonyl]-N';-hydroxyethanimidamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.