missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Isocyanatophenyl)pyrimidine, 90%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 89.5 |
CAS Max % | 100.0 |
Assay Percent Range | 90% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 197.2 |
Percent Purity | 90% |
Chemical Name or Material | 2-(4-Isocyanatophenyl)pyrimidine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction