missing translation for 'onlineSavingsMsg'
Learn More
2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid, 97%, Thermo Scientific™ Product Code.: 10394652

2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid, 97%, Thermo Scientific™

Product Code. 10394652
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10394652

Brand: Thermo Scientific Maybridge BTB09316DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 77978-73-5
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00173859
InChI Key WLKUVVIAHUXPSB-UHFFFAOYSA-N
Synonym 2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3-yl acetic acid, 4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 1h-pyrrole-3-aceticacid, 2,3-dihydro-4-methoxycarbonyl-5-methyl-2-oxo, maybridge1_002247, 3-methoxycarbonyl-2-methyl-5-oxo-2-pyrrolin-4-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3yl acetic acid
PubChem CID 2774998
IUPAC Name 2-(4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl)acetic acid
SMILES CC1=C(C(C(=O)N1)CC(=O)O)C(=O)OC

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 213.19
Percent Purity 97%
Chemical Name or Material 2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2, 3-dihydro-1H-pyrrol-3-yl]acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.