missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-methoxyphenoxy)ethanamine hydrochloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 203.67 |
Chemical Name or Material | 2-(4-methoxyphenoxy)ethanamine hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'