Learn More
2,4-Pentanedione, 99+%, Thermo Scientific Chemicals
£21.40 - £355.00
Chemical Identifiers
CAS | 123-54-6 |
---|---|
Molecular Formula | C5H8O2 |
Molecular Weight (g/mol) | 100.12 |
MDL Number | MFCD00008787 |
InChI Key | YRKCREAYFQTBPV-UHFFFAOYSA-N |
Synonym | 2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone |
PubChem CID | 31261 |
ChEBI | CHEBI:14750 |
IUPAC Name | pentane-2,4-dione |
SMILES | CC(=O)CC(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10224990
|
Thermo Scientific Acros
|
5 g |
£21.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10604403
|
Thermo Scientific Acros
|
250 g |
£21.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10128412
|
Thermo Scientific Acros
|
1 kg |
£60.80
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10493825
|
Thermo Scientific Acros
|
2.5 kg |
£120.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11954821
|
Thermo Scientific Acros
|
10 kg |
£355.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
123-54-6 | |
100.12 | |
YRKCREAYFQTBPV-UHFFFAOYSA-N | |
31261 | |
pentane-2,4-dione |
C5H8O2 | |
MFCD00008787 | |
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone | |
CHEBI:14750 | |
CC(=O)CC(C)=O |
Specifications
123-54-6 | |
0.9750g/mL | |
35.5°C | |
99% min. (GC) | |
C5H8O2 | |
1.4510 to 1.4530 | |
01,777 | |
15,76 | |
2,4-pentanedione, acetylacetone, 2,4-pentadione, diacetylmethane, acetoacetone, acac, 2,4-dioxopentane, pentanedione, pentan-2,4-dione, acetyl acetone | |
YRKCREAYFQTBPV-UHFFFAOYSA-N | |
pentane-2,4-dione | |
31261 | |
0.7445 mPa.s (20°C) | |
99+% | |
2, 4-Pentanedione |
-23.0°C | |
133.0°C to 135.0°C | |
Authentic | |
Glass bottle | |
CH3COCH2COCH3 | |
MFCD00008787 | |
01,1 | |
0.975 | |
Solubility in water: 160g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in alcohol, chloroform, ether, benzene, and acetone | |
CC(=O)CC(C)=O | |
100.12 | |
CHEBI:14750 | |
100.12 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 204-634-
- 2,4-Pentanedione
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Flammable liquid Category 3
Hazard Statement
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P311-Call a POISON CENTER/doctor/ .
RUO – Research Use Only