missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Pentanedione, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.
Solubility
It is readily miscible with water.
Notes
Incompatible with isocyanates, aldehydes, cyanides, peroxides, and anhydrides.
2,4-Pentanedione is used as a precursor to acetylacetonate, a common bidentate and a raw material for the synthesis of heterocyclic compounds. It is used in pharmaceuticals, organic intermediates for solvents, analytical reagents, tungsten and molybdenum in aluminum extraction agent.
Solubility
It is readily miscible with water.
Notes
Incompatible with isocyanates, aldehydes, cyanides, peroxides, and anhydrides.
Specifications
Specifications
Melting Point | -23°C |
Density | 0.973 |
Boiling Point | 140°C to 141°C |
Flash Point | 34°C (93°F) |
Assay Percent Range | 99% |
Refractive Index | 1.452 |
Quantity | 2500 mL |
UN Number | UN2310 |
Beilstein | 741937 |
Merck Index | 14,81 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'