missing translation for 'onlineSavingsMsg'
Learn More

2-(5-Bromo-2-thienyl)pyridine, 97%, Thermo Scientific™

Product Code. 10232492
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 123784-07-6
Molecular Formula C9H6BrNS
Molecular Weight (g/mol) 240.12
MDL Number MFCD00120466
InChI Key AKNKWLQVFAYRRO-UHFFFAOYSA-N
Synonym 2-5-bromo-2-thienyl pyridine, 2-5-bromothiophen-2-yl pyridine, 2-5-bromothien-2-yl pyridine, pyridine,2-5-bromo-2-thienyl, acmc-20mqrz, 5-bromo-2-2-pyridyl thiophene, 2-5-bromo-2-thiophenyl pyridine, 2-bromo-5-2-pyridinyl thiophene, 2-5-bromanylthiophen-2-yl pyridine, 2-5-bromo-thiophen-2-yl-pyridine
PubChem CID 736483
IUPAC Name 2-(5-bromothiophen-2-yl)pyridine
SMILES BrC1=CC=C(S1)C1=CC=CC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 240.12
Percent Purity 95%
Chemical Name or Material 2-(5-Bromo-2-thienyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'