missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 240.71 |
Percent Purity | 95% |
Chemical Name or Material | 2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'