missing translation for 'onlineSavingsMsg'
Learn More

2,5-Di-tert-butylhydroquinone, 99%, Thermo Scientific Chemicals

Product Code. 10404555
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-58-4
Molecular Formula C14H22O2
Molecular Weight (g/mol) 222.33
MDL Number MFCD00008825
InChI Key JZODKRWQWUWGCD-UHFFFAOYSA-N
Synonym 2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
PubChem CID 2374
ChEBI CHEBI:41094
IUPAC Name 2,5-ditert-butylbenzene-1,4-diol
SMILES CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 212°C to 218°C
Color Beige to Beige-Pink
Density 216g/mL
Flash Point 216°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 2.5 kg
Solubility Information Solubility in water: insoluble in water. Other solubilities: insoluble in aqueous alkali,soluble in acetone,alcohol and benzene,practically insoluble in toluene and hexane
Formula Weight 222.33
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 5-Di-tert-butylhydroquinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'