missing translation for 'onlineSavingsMsg'
Learn More

2,5-Diaminopyridine, 98+%, ACROS Organics™

Product Code. 10282902
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4318-76-7
Molecular Formula C5H7N3
Molecular Weight (g/mol) 109.13
InChI Key MIROPXUFDXCYLG-UHFFFAOYSA-N
Synonym 2,5-diaminopyridine, 2,5-pyridinediamine, pyridine, 2,5-diamino, diaminopyridine van, unii-h6olz060hi, h6olz060hi, da1, pyridine,5-diamino, 2,5-diamino pyridine, 2,5-diamino-pyridine
PubChem CID 20314
IUPAC Name pyridine-2,5-diamine
SMILES C1=CC(=NC=C1N)N

Specifications

Boiling Point 180.0°C (12.0 mmHg)
Assay Percent Range 98% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 109.13
Percent Purity 97%
Chemical Name or Material 2, 5-Diaminopyridine
compliance-icons
Product Identifier
  • 2,5-Diaminopyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'