missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dibromohydroquinone, 97%, Thermo Scientific Chemicals

£25.00 - £166.83

Chemical Identifiers

CAS 14753-51-6
Molecular Formula C6H4Br2O2
Molecular Weight (g/mol) 267.904
MDL Number MFCD00192664
InChI Key VALXCIRMSIFPFN-UHFFFAOYSA-N
PubChem CID 280945
IUPAC Name 2,5-dibromobenzene-1,4-diol
SMILES C1=C(C(=CC(=C1Br)O)Br)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15451698
View Documents
Thermo Scientific Alfa Aesar
H55453.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15461698
View Documents
Thermo Scientific Alfa Aesar
H55453.06
5 g
£46.13
5g
Please to purchase this item. Need a web account? Register with us today!
15471698
View Documents
Thermo Scientific Alfa Aesar
H55453.14
25 g
£166.83
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is an important pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important pharmaceutical intermediate.

Solubility
Soluble in methanol

Notes
Storage: Keep container tightly closed. Store in a cool, dark and well-ventilated place and away from oxidizing agents.
Specifications

Chemical Identifiers

14753-51-6
267.904
VALXCIRMSIFPFN-UHFFFAOYSA-N
2,5-dibromobenzene-1,4-diol
C6H4Br2O2
MFCD00192664
280945
C1=C(C(=CC(=C1Br)O)Br)O

Specifications

14753-51-6
C6H4Br2O2
2,5-dibromohydroquinone, 2,5-dibromo-1,4-benzenediol, 2,5-dibromohydroquine, 1,4-benzenediol,2,5-dibromo, 2,5-dibromo-benzene-1,4-diol, 1,4-benzenediol, 2,5-dibromo, 2,5-dibromo-1,4-dihydroxybenzene
VALXCIRMSIFPFN-UHFFFAOYSA-N
2,5-dibromobenzene-1,4-diol
280945
97%
191°C to 194°C
MFCD00192664
Soluble in methanol
C1=C(C(=CC(=C1Br)O)Br)O
267.904
267.9
2,5-Dibromohydroquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only