missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloro-4,6-dimethylnicotinonitrile, ≥95%, Thermo Scientific™

Product Code. 10673793
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
missing translation for 'unitSize'
10g
1g
This item is not returnable. View return policy

Product Code. 10673793

missing translation for 'mfr': Thermo Scientific Maybridge RF00821DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 91591-63-8
Molecular Formula C8H6Cl2N2
Molecular Weight (g/mol) 201.05
MDL Number MFCD00052631
InChI Key UCGWYTUBYASPFG-UHFFFAOYSA-N
PubChem CID 334852
IUPAC Name 2,5-dichloro-4,6-dimethylpyridine-3-carbonitrile
SMILES CC1=C(C(=NC(=C1Cl)C)Cl)C#N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2,5-dichloro-4,6-dimethylnicotinonitrile, 3-pyridinecarbonitrile, 2,5-dichloro-4,6-dimethyl, 2,5-dichloro-4,6-dimethyl-nicotinonitrile, pubchem20434, ksc669q6f, 2,5-dichloro-3-cyano-4,6-dimethylpyridine, 2,5-dichloro-4,6-dimethyl-3-pyridyl cyanide, 3-pyridinecarbonitrile,5-dichloro-4,6-dimethyl, 2,5-dichloro-4,6-dimethyl-pyridine-3-carbonitrile, 3-pyridinecarbonitrile,2,5-dichloro-4,6-dimethyl
Formula Weight 201.06
Percent Purity 95%
Chemical Name or Material 2, 5-Dichloro-4, 6-dimethylnicotinonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.