missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichloroaniline, 99%, Thermo Scientific Chemicals

£41.51 - £252.00

Chemical Identifiers

CAS 95-82-9
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.01
MDL Number MFCD00007667
InChI Key AVYGCQXNNJPXSS-UHFFFAOYSA-N
Synonym 1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
PubChem CID 7262
ChEBI CHEBI:34245
IUPAC Name 2,5-dichloroaniline
SMILES NC1=CC(Cl)=CC=C1Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11414080
View Documents
Thermo Scientific Alfa Aesar
A11151.30
250 g
£41.51
250g
Please to purchase this item. Need a web account? Register with us today!
11419073
View Documents
Thermo Scientific Alfa Aesar
A11151.36
500 g
£64.70
500g
Please to purchase this item. Need a web account? Register with us today!
11429073
View Documents
Thermo Scientific Alfa Aesar
A11151.0E
2500 g
£252.00
2500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

Solubility
Soluble in dilute hydrochloric acid.

Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

95-82-9
162.01
AVYGCQXNNJPXSS-UHFFFAOYSA-N
7262
2,5-dichloroaniline
C6H5Cl2N
MFCD00007667
1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
CHEBI:34245
NC1=CC(Cl)=CC=C1Cl

Specifications

95-82-9
250°C to 251°C
C6H5Cl2N
UN3442
1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
NC1=CC(Cl)=CC=C1Cl
162.01
CHEBI:34245
99%
48°C to 52°C
139°C (282°F)
MFCD00007667
1447438
AVYGCQXNNJPXSS-UHFFFAOYSA-N
2,5-dichloroaniline
7262
162.02
2,5-Dichloroaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P261-P301+P310-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID

missing translation for 'rtecsNumber' : BX2610000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only