missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorophenyl isocyanate, 97%, Thermo Scientific™

Product Code. 11302247
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5392-82-5
Molecular Formula C7H3Cl2NO
Molecular Weight (g/mol) 188.01
MDL Number MFCD00002002
InChI Key PEQMJVGRHNZPAM-UHFFFAOYSA-N
Synonym 2,5-dichlorophenyl isocyanate, benzene, 1,4-dichloro-2-isocyanato, 2,5-dichlorophenylisocyanate, isocyanic acid, 2,5-dichlorophenyl ester, isocyanic acid 2,5-dichlorophenyl ester, acmc-209laz, 2,5-dichlorobenzenisocyanate, benzene,4-dichloro-2-isocyanato, #, isocyanic acid,5-dichlorophenyl ester
PubChem CID 79360
IUPAC Name 1,4-dichloro-2-isocyanatobenzene
SMILES ClC1=CC=C(Cl)C(=C1)N=C=O

Specifications

Melting Point 28°C to 31°C
Boiling Point 120°C to 122°C (20 mmHg)
Flash Point >110°C (230°F)
Odor Irritating
Quantity 5 g
UN Number UN2250
Beilstein 388960
Sensitivity Moisture sensitive
Formula Weight 188.01
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorophenyl isocyanate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'